missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PrPC Antibody (S03-2E2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19967-100ul
This item is not returnable.
View return policy
Description
PrPC Monoclonal antibody specifically detects PrPC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| PrPC | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry 1:20, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
| A synthetic peptide of human PrPC (Uniprot # P04156) | |
| 100 μg | |
| Neuroscience, Stem Cell Markers | |
| 5621 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S03-2E2 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction