missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRPF3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15743471 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15743471 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15743471 Supplier Invitrogen™ Supplier No. PA555550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WWDSYIIPNG FDLTEENPKR EDYFGITNLV EHPAQLNPPV DNDTPVTLGV YLTKKEQKKL RRQTRREAQK ELQEKVRLGL MPPPEPKVRI SNLMRVLGTE AVQDPTK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs. The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF3
Gene Accession No. O43395
Gene Alias 3632413F13Rik; HPRP3; HPRP3P; MGC137653 protein; pre-mRNA processing factor 3; pre-mRNA-splicing factor 3; PRP3; PRP3 pre-mRNA processing factor 3 homolog; PRP3 pre-mRNA processing factor 3 homolog (yeast); Prp3p; PRPF3; RP18; SNRNP90; U4/U6 small nuclear ribonucleoprotein Prp3; U4/U6 snRNP 90 kDa protein; U4/U6-associated RNA splicing factor
Gene Symbols PRPF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRPF3. Recombinant protein control fragment (Product #RP-94114).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9129
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.