missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF38A Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37697AF350
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PRPF38A Polyclonal antibody specifically detects PRPF38A in Human, Mouse samples. It is validated for Western Blot
Tekniske data
| PRPF38A | |
| Polyclonal | |
| Western Blot | |
| FLJ14936, MGC3320, pre-mRNA-splicing factor 38A, Prp38, PRP38 pre-mRNA processing factor 38 (yeast) domain containing A, RP5-965L7.1 | |
| This antibody maps to a region between residue 262 to 312 of human Pre-mRNA Processing Factor 38A using the numbering given in entry NP_116253.2 (GeneID 84950). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84950 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion