missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF38A Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37697H
This item is not returnable.
View return policy
Description
PRPF38A Polyclonal antibody specifically detects PRPF38A in Human, Mouse samples. It is validated for Western Blot
Specifications
| PRPF38A | |
| Polyclonal | |
| Western Blot | |
| FLJ14936, MGC3320, pre-mRNA-splicing factor 38A, Prp38, PRP38 pre-mRNA processing factor 38 (yeast) domain containing A, RP5-965L7.1 | |
| This antibody maps to a region between residue 262 to 312 of human Pre-mRNA Processing Factor 38A using the numbering given in entry NP_116253.2 (GeneID 84950). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84950 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction