missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF4B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93540-0.1ml
This item is not returnable.
View return policy
Description
PRPF4B Polyclonal antibody specifically detects PRPF4B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRPF4B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dJ1013A10.1, dJ1013A10.1 (PRP4 protein kinase homolog), KIAA0536EC 2.7.11.1, PR4H, protein-serine/threonine kinase, PRP4 kinase, PRP4 pre-mRNA processing factor 4 homolog B (yeast), PRP4 pre-mRNA-processing factor 4 homolog, PRP4H, PRP4K, PRP4Prp4, serine/threonine-protein kinase PRP4 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 550-760 of human PRPF4B (NP_003904.3). QKYKYLAEDSNMSVPSEPSSPQSSTRTRSPSPDDILERVAADVKEYERENVDTFEASVKAKHNLMTVEQNNGSSQKKLLAPDMFTESDDMFAAYFDSARLRAAGIGKDFKENPNLRDNWTDAEGYYRVNIGEVLDKRYNVYGYTGQGVFSNVVRARDNARANQEVAVKIIRNNELMQKTGLKELEFLKKLNDADPDDKFHCLRLFRHFYHK | |
| 0.1 mL | |
| Protein Kinase | |
| 8899 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction