missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46213
This item is not returnable.
View return policy
Description
PRPF6 Polyclonal specifically detects PRPF6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| PRPF6 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| PRPF6 | |
| The immunogen for this product maps to a region between residue 150 and 200 of human Pre-mRNA Processing Factor 6 using the numbering given in entry NP_036601.2 (GeneID 24148). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate | |
| androgen receptor N-terminal domain transactivating protein-1, ANT-1, bB152O15.1, C20orf14, chromosome 20 open reading frame 14, hPrp6, p102 U5 small nuclear ribonucleoprotein particle-binding protein, pre-mRNA-processing factor 6, Prp6, PRP6 homolog, PRP6 pre-mRNA processing factor 6 homolog (S. cerevisiae), PRP6 pre-mRNA processing factor 6 homolog (yeast), putative mitochondrial outer membrane protein import receptor, TOM, U5 snRNP-associated 102 kDa protein, U5-102 kDa protein, U5-102K | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 24148 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction