missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPH Monoclonal Antibody (OTI7E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806226
This item is not returnable.
View return policy
Description
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.Specifications
| PRPH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41219 | |
| PRPH | |
| Full length human recombit protein of human PRPH produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI7E4 | |
| Unconjugated | |
| PRPH | |
| NEF4, PRPH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5630 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction