missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPK Antibody (OTI5B2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73675CL1
This item is not returnable.
View return policy
Description
PRPK Monoclonal antibody specifically detects PRPK in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PRPK | |
| Monoclonal | |
| CoraFluor 1 | |
| BUD32, C20orf64, chromosome 20 open reading frame 64, dJ101A2, dJ101A2.2, EC 2.7.11.1, Nori-2, Nori-2p, p53-related protein kinase, PRPK, TP53 regulating kinase, TP53-regulating kinase | |
| Full length human recombinant protein of human TP53RK (NP_291028) produced in E.coli. | |
| 0.1 mL | |
| Protein Kinase | |
| 112858 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI5B2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction