missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93816-0.02ml
This item is not returnable.
View return policy
Description
PRPS1 Polyclonal antibody specifically detects PRPS1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRPS1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CMTX5ARTS, deafness, X-linked 2, perceptive, congenital, DFN2, DFNX1KIAA0967, dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1), EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase I, phosphoribosyl pyrophosphate synthetase 1, PPRibP, PRSI, PRS-I, ribose-phosphate diphosphokinase 1, ribose-phosphate pyrophosphokinase 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-318 of human PRPS1 (NP_002755.1). NEVDRMVLVGDVKDRVAILVDDMADTCGTICHAADKLLSAGATRVYAILTHGIFSGPAISRINNACFEAVVVTNTIPQEDKMKHCSKIQVIDISMILAEAIRRTHNGESVSYLFSHVPL | |
| 0.02 mL | |
| Apoptosis, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 5631 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction