missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPS1L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116324
This item is not returnable.
View return policy
Description
PRPS1L1 Polyclonal Antibody for Western Blot
Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.
Specifications
| PRPS1L1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P21108 | |
| PRPS1L1 | |
| KLH conjugated synthetic peptide between 77-106 amino acids from the N-terminal region of human PRPS1L1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PRPS1L1 | |
| 1700011K15Rik; hCG_1643027; phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosyl pyrophosphate synthetase 1-like 1; phosphoribosylpyrophosphate synthetase subunit III; PRPS1; Prps1l1; PRPS1-like 1; PRPS3; PRPSL; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 221823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction