missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPSAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590638
This item is not returnable.
View return policy
Description
Immunogen sequence: MNAARTGYRV FSANSTAACT ELAKRITERL GAELGKSVVY QETNGETRVE IKESVRGQDI FIIQTIPRDV NTAVMELLIM A; Positive Samples: HeLa, Mouse heart.
Seems to play a negative regulatory role in 5-phosphoribose 1-diphosphate synthesis. Target protein binds catalytic subunits of the ribose-phosphate diphosphokinase enzyme complex; may act as a negative regulator of enzyme activity.
Specifications
| PRPSAP1 | |
| Polyclonal | |
| Unconjugated | |
| PRPSAP1 | |
| 39 kDa phosphoribosypyrophosphate synthase-associated protein; 39 kDa phosphoribosypyrophosphate synthetase-associated protein; 5730409F23Rik; hypothetical protein LOC431746; kpra; PAP39; Phosphoribosyl pyrophosphate synthase-associated protein 1; phosphoribosyl pyrophosphate synthetase associated protein 1; phosphoribosyl pyrophosphate synthetase-associated protein 1; phosphoribosylpyrophosphate synthetase-associated protein (39 kDa); PRPP synthase-associated protein 1; PRPP synthetase-associated protein 1; Prpsap1; zgc:91845 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5635, 67763 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q14558, Q9D0M1 | |
| PRPSAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-110 of human PRPSAP1 (NP_0027572). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction