missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPSAP2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-97949F
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PRPSAP2 Polyclonal specifically detects PRPSAP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| PRPSAP2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| MGC117304, MGC126719, MGC126721, PAP4141 kDa phosphoribosypyrophosphate synthetase-associated protein, phosphoribosyl pyrophosphate synthase-associated protein 2, phosphoribosyl pyrophosphate synthetase-associated protein 2, PRPP synthase-associated protein 2 | |
| Produced in rabbits immunized with E. coli-derived Human PRPSAP2 fragment. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 5636 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido