missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Sheep IgG F(ab')2 Secondary Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72808PCP
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| PBS | |
| Sheep IgG F(ab')2 fragment | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| PerCP | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction