missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRPSAP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15782461 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15782461 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782461 Supplier Invitrogen™ Supplier No. PA554533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VTPPELETKM NITKGGLVLF SANSNSSCME LSKKIAERLG VEMGKVQVYQ EPNRETRVQI QES Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPSAP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prpsap2
Gene Accession No. O60256
Gene Alias 41 kDa phosphoribosypyrophosphate synthetase-associated protein; A230054F23Rik; I79_001737; Pap41; phosphoribosyl pyrophosphate synthase-associated protein 2; phosphoribosyl pyrophosphate synthetase associated protein 2; phosphoribosyl pyrophosphate synthetase-associated protein 2; Phosphoribosyl pyrophosphate synthetase-associated protein 2-like protein; PRPP synthase-associated protein 2; PRPP synthetase-associated protein 2; Prpsap2; prpsap2 protein; wu:fi14g04; zgc:55477; zgc:85610
Gene Symbols Prpsap2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRPSAP2. Recombinant protein control fragment (Product #RP-93271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5636
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.