missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19967
This item is not returnable.
View return policy
Description
PRR11 Polyclonal specifically detects PRR11 in Human samples. It is validated for Western Blot.
Specifications
| PRR11 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| FLJ11029, proline rich 11, proline-rich protein 11, transcription repressor of MHCII | |
| Rabbit | |
| 40 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: mouse (80%), Rat (80%), cow (88%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96HE9 | |
| PRR11 | |
| Recombinant protein encompassing a sequence within the center region of human PRR11. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 55771 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction