missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR11 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800449S
This item is not returnable.
View return policy
Description
PRR11 Monoclonal antibody specifically detects PRR11 in Canine, Human samples. It is validated for Western BlotSpecifications
| PRR11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NAM6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 106559239, 201516 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A4 | |
| Unconjugated | |
| ZSCAN4 | |
| B930067F20Rik; proline rich 11; proline-rich protein 11; PRR11; transcription repressor of MHCII | |
| Human recombinant protein fragment corresponding to amino acids 164-433 of human ZSCAN4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction