missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRR11 Monoclonal Antibody (OTI1C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526460
This item is not returnable.
View return policy
Description
PRR11 Monoclonal Antibody for Western Blot, IHC (P)
Plays a critical role in cell cycle progression.
Specifications
| PRR11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96HE9 | |
| PRR11 | |
| Human recombinant protein fragment corresponding to amino acids 125-360 of PRR11 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C1 | |
| Unconjugated | |
| PRR11 | |
| B930067F20Rik; proline rich 11; proline-rich protein 11; PRR11; transcription repressor of MHCII | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55771 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction