Learn More
PRR13 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14101-1-AP-150UL
 View more versions of this product
Product Code. 16882584
  • € 410.00 / 150µL
Description
Negatively regulates TSP1 expression at the level of transcription. This down-regulation was shown to reduce taxane-induced apoptosis.
Specifications
Specifications
PRR13
Polyclonal
Unconjugated
PRR13
proline rich 13, Proline rich protein 13, PRR13, Taxane resistance protein, TXR1
Rabbit
Antigen Affinity Chromatography
RUO
54458
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NZ81
PRR13
PRR13 Fusion Protein Ag5280
150 μL