missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR20A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46857
This item is not returnable.
View return policy
Description
PRR20A Polyclonal antibody specifically detects PRR20A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PRR20A | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P86481 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 122183 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Proline-rich protein 20A;Proline-rich protein 20B;Proline-rich protein 20C;Proline-rich protein 20D;Proline-rich protein 20E;PRR20;PRR20B;PRR20C;PRR20D;PRR20E | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MEEPRPSKRLRSMAPNQASGGPPPEPGCCVADPEGSVEADGPAQPAQPAKPIAYVKPFRRQP | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction