missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRR20A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA571242

Product Code. 15829653

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This target displays homology in the following species: Human: 100%.

This gene is one of five identical loci in a cluster on chromosome 13q21.1. The predicted protein is proline-rich and contains several dopamine D4 receptor signatures and PRINTS domains.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRR20A
Polyclonal
Unconjugated
PRR20A
proline rich 20; proline rich 20A; proline-rich protein 20; proline-rich protein 20A; Proline-rich protein 20B; Proline-rich protein 20C; Proline-rich protein 20D; Proline-rich protein 20E; PRR20; PRR20A; PRR20B; PRR20C; PRR20D; PRR20E
Rabbit
Affinity Chromatography
RUO
122183
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P86496
PRR20A
synthetic peptide directed towards the N-terminal region of human PRR20A.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.