missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR7 Antibody (TRAP3/10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30424V
This item is not returnable.
View return policy
Description
PRR7 Monoclonal specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PRR7 | |
| Monoclonal | |
| DyLight 405 | |
| MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 80758 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| TRAP3/10 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| PRR7 | |
| Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253) | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7 TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction