missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRC2B Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98065C
This item is not returnable.
View return policy
Description
PRRC2B Polyclonal specifically detects PRRC2B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PRRC2B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BAT2L, DKFZp781F05101, HLA-B associated transcript 2-like 1, HLA-B-associated transcript 2-like 1, LQFBS-1, MGC10526, proline-rich coiled-coil 2B, proline-rich coiled-coil protein 2B, protein BAT2-like 1, protein PRRC2B | |
| Produced in rabbits immunized with E. coli-derived Human PRRC2B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 84726 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction