missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRRG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569302
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.
This gene encodes a vitamin K-dependent, gamma-carboxyglutamic acid (Gla)-containing, single-pass transmembrane protein. This protein contains a Gla domain at the N-terminus, preceded by a propeptide sequence required for post-translational gamma-carboxylation of specific glutamic acid residues by a vitamin K-dependent gamma-carboxylase. The C-terminus is proline-rich containing PPXY and PXXP motifs found in a variety of signaling and cytoskeletal proteins. This gene is highly expressed in the spinal cord. Several alternatively spliced transcript variants have been found for this gene.
Specifications
| PRRG1 | |
| Polyclonal | |
| Unconjugated | |
| PRRG1 | |
| 2010007L08Rik; ortholog of proline rich Gla (G-carboxyglutamic acid) 1 PRRG1; PRGP1; proline rich and Gla domain 1; proline rich Gla (G-carboxyglutamic acid) 1; Proline-rich gamma-carboxyglutamic acid protein 1; proline-rich Gla (G-carboxyglutamic acid) polypeptide 1; Proline-rich Gla protein 1; PRRG1; RGD1561320; TMG1; Transmembrane gamma-carboxyglutamic acid protein 1; transmembrane gamma-carboxyglutamic acid protein 1-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5638 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O14668 | |
| PRRG1 | |
| synthetic peptide directed towards the N terminal region of human PRRG1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction