missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRRG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115801
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PRRG4.
PRRG4 gene ontology annotations related to this gene include calcium ion binding.
Specifications
| PRRG4 | |
| Polyclonal | |
| Unconjugated | |
| PRRG4 | |
| 9930111I18Rik; hypothetical protein LOC550330; PRGP4; proline rich and Gla domain 4; proline rich Gla (G-carboxyglutamic acid) 4 (transmembrane); proline rich Gla 4 (transmembrane) isoform 1; proline rich Gla 4 (transmembrane) isoform 2; proline rich Gla 4 (transmembrane) isoform 3; proline-rich gamma-carboxyglutamic acid protein 4; Proline-rich Gla protein 4; PRRG4; RGD1560542; TMG4; Transmembrane gamma-carboxyglutamic acid protein 4; zgc:110234 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 228413, 499847, 79056 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8BGN6, Q9BZD6 | |
| PRRG4 | |
| A synthesized peptide derived from human PRRG4(Accession Q9BZD6), corresponding to amino acid residues E81-F131. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction