missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRRS Virus M Protein (Strain: VR-2332) Recombinant Rabbit Monoclonal Antibody (HL2619)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556848
This item is not returnable.
View return policy
Description
PRRS Virus M Protein (Strain: VR-2332) Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Porcine reproductive and respiratory syndrome virus (PRRSV) is a highly infectious pathogen that causes severe diseases in pigs and great economic losses to the swine industry worldwide. GP5 and M are the major envelope proteins encoded by PRRSV.
Specifications
| PRRS Virus M Protein (Strain: VR-2332) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2619 | |
| Unconjugated | |
| Porcine reproductive and respiratory syndromevirus | |
| Recombinant fragment of PRRS virus M protein (Strain: VR-2332). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction