missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRRS Virus Nucleocapsid Protein Recombinant Rabbit Monoclonal Antibody (HL1901)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA547214

Product Code. 17849525

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Betaarterivirus suid 1, formerly (PRRS, Porcine reproductive and respiratory syndrome virus), is a virus that causes a disease of pigs, called porcine reproductive and respiratory syndrome, also known as blue-ear pig disease. The Nucleocapsid protein is an internal structure of the virus that comprised of the association of the N protein and the RNA genome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRRS Virus Nucleocapsid Protein
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Western Blot, Immunocytochemistry
HL1901
Unconjugated
Porcine reproductive and respiratory syndrome virus ORF7; PRRSV ORF7
Full length PRRS virus Nucleocapsid protein.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.