missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRT2 Antibody (CL4233), Novus Biologicals™
Mouse Monoclonal Antibody
€ 280.00
Specifications
| Antigen | PRRT2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
PRRT2 Monoclonal specifically detects PRRT2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PRRT2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| DKFZp547J199, FLJ25513, IFITMD1, interferon induced transmembrane protein domain containing 1, proline-rich transmembrane protein 2 | |
| PRRT2 | |
| IgG1 | |
| Protein A purified |
| Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| Unconjugated | |
| Mouse | |
| Human | |
| 112476 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PKPALQPELPTQEDPTPEILSESVGEKQENGAVVPLQAGDGEEGPAPEPHSPPSKKSPPANGAPPRVLQQLVEEDRMRRAHSGHPGSPRGSLSRHPSSQLAGPGVEGGEGTQKPRDY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title