missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRX1 Antibody (OTI1E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45407
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
PRRX1 Monoclonal specifically detects PRRX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PRRX1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1E10 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| PRRX1 | |
| Full length human recombinant protein of human PRRX1 (NP_073207) produced in E.coli. | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 5396 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction