missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRRX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106700
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PRRX1.
The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and the control of gene expression. PRX1 (paired related homeobox 1), also known as PRRX1, PMX1 or PHOX1, is a 245 amino acid protein that contains one OAR domain and one homeobox DNA-binding domain and belongs to the paired homeobox family. Localized to the nucleus, PRX1 functions as a transcriptional co-activator that enhances the DNA-binding activity of serum response factor (SRF), thereby mediating the induction of SRF-dependent gene expression by growth and differentiation factors. Additionally, PRX1 regulates the transcriptional activities of creatine kinase-M (muscle), thereby playing a role in the establishment of mesodermal muscle types. PRX1 exists as two alternatively spliced isoforms, designated PMX1-A and PMX1-B.
Specifications
| PRRX1 | |
| Polyclonal | |
| Unconjugated | |
| PRRX1 | |
| A230024N07Rik; AA755424; AGOTC; AI385634; AI843499; Homeobox protein K-2; homeobox protein mHox; Homeobox protein PHOX1; K-2; mHox; paired mesoderm homeobox 1; paired mesoderm homeobox 1 isoform pmx-1b; paired mesoderm homeobox protein 1; paired related homeobox 1; paired-related homeobox protein 1; PHOX1; Pmx; Pmx1; Prrx1; Prx1; PRX-1; rHox | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18933, 266813, 5396 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54821, P63013, P63014 | |
| PRRX1 | |
| A synthesized peptide derived from human PRRX1(Accession P54821), corresponding to amino acid residues A53-S103. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction