missing translation for 'onlineSavingsMsg'
Learn More

PRRX2, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16156996
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16156996 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16156996 Supplier Abnova Supplier No. H00051450D01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence: MDSAAAAFALDKPALGPGPPPPPPALGPGDCAQARKNFSVSHLLDLEEVAAAGRLAARPGARAEAREGAAREPSGGSSGSEAAPQDGECPSPGRGSAAKRKKKQRRNRTTFNSSQLQALERVFERTHYPDAFVREELARRVNLSEARVQVWFQNRRAKFRRNERAMLASRSASLLKSYSQEAAIEQPVAPRPTALSPDYLSWTASSPYSTVPPYSPGSSGPATPGVNMANSIASLRLKAKEFSLHHSQVPTVN

Specifications

Antigen PRRX2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation No additive
Gene paired related homeobox 2
Gene Accession No. NM_016307.3
Gene Alias MGC19843/PMX2/PRX2
Gene Symbols PRRX2
Host Species Rabbit
Immunogen PRRX2 (NP_057391.1, 1 a.a. ∼ 253 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51450
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.