missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRTFDC1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76358F
This item is not returnable.
View return policy
Description
PRTFDC1 Polyclonal antibody specifically detects PRTFDC1 in Human, Rat samples. It is validated for ELISA
Specifications
| PRTFDC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA | |
| FITC | |
| EC 2.4.2.8, FLJ11888, HHGPEC 2.4.2, phosphoribosyl transferase domain containing 1, phosphoribosyltransferase domain-containing protein 1 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the center of human PRTFDC1. The immunogen is located within amino acids 90 - 140 of PRTFDC1. Amino Acid Squence: MKVDFIRLKSYRND | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 56952 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction