missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRTFDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19972
This item is not returnable.
View return policy
Description
PRTFDC1 Polyclonal specifically detects PRTFDC1 in Human samples. It is validated for Western Blot.
Specifications
| PRTFDC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EC 2.4.2.8, FLJ11888, HHGPEC 2.4.2, phosphoribosyl transferase domain containing 1, phosphoribosyltransferase domain-containing protein 1 | |
| Rabbit | |
| 26 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 56952 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NRG1 | |
| PRTFDC1 | |
| Recombinant protein encompassing a sequence within the center region of human PRTFDC1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Chicken (83%), Rat (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction