missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRTN3 Monoclonal Antibody (OTI1D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807292S
This item is not returnable.
View return policy
Description
PRTN3 Monoclonal antibody specifically detects PRTN3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PRTN3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14763 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8795 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D10 | |
| Unconjugated | |
| TNFRSF10B | |
| ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen | |
| Human recombinant protein fragment corresponding to amino acids 203-411 of human TNFRSF10B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction