Learn More
PRTN3 Mouse anti-Human, Clone: OTI9C2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807348
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Polymorphonuclear leukocyte serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) and causes emphysema when administered by tracheal insufflation to hamsters.Specifications
| PRTN3 | |
| Monoclonal | |
| Unconjugated | |
| PRTN3 | |
| ACPA, AGP7, C-ANCA, CANCA, MBN, MBT, NP-4, NP4, P29, PR-3, PR3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5657 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9C2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P24158 | |
| PRTN3 | |
| Human recombit protein fragment corresponding to amino acids 28-248 of human PRTN3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.