missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PRUNE Antibody (1C11), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27951

Product Code. 30044722

  • € 917.00 / 0.05mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PRUNE Monoclonal antibody specifically detects PRUNE in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRUNE
Monoclonal
0.5 mg/mL
Western Blot, ELISA, Immunohistochemistry-Paraffin 5 μg/mL
Q86TP1
Mouse
Protein A purified
RUO
58497
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG2a κ
ELISA, Western Blot, Immunohistochemistry (Paraffin)
1C11
Unconjugated
PBS (pH 7.4)
BMCC1, DRES17, DRES-17EC 3.6.1.1, Drosophila-related expressed sequence 17, hPrune, HTCD37, KIAA0367, protein prune homolog, prune homolog (Drosophila), PRUNE2
Recombinant protein
0.05 mg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.