missing translation for 'onlineSavingsMsg'
Learn More

PRX-08066, MedChemExpress

Product Code. 16439137
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16439137

Brand: Medchem Express HY15472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PRX-08066 is a selective 5-hydroxytryptamine receptor 2B (5-HT2BR, IC50= 3.4 nM) antagonist that causes selective vasodilation of pulmonary arteries.

PRX-08066 is a selective 5-hydroxytryptamine receptor 2B (5-HT2BR, IC50= 3.4 nM) antagonist that causes selective vasodilation of pulmonary arteries.

Specifications

Chemical Name or Material PRX-08066
CAS 866206-54-4
Color off White
Purity Grade Notes Research
Molecular Formula C19H17ClFN5S
Quantity 5 mg
Solubility Information DMSO : 7 mg/mL (17.42 mM; Need ultrasonic)
SMILES N#CC1=CC(CN2CCC(NC3=C(C=C(Cl)S4)C4=NC=N3)CC2)=CC=C1F
Molecular Weight (g/mol) 401.89
Formula Weight 401.89
Percent Purity 97.62%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.