missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PS 1145 dihydrochloride

Product Code. 18760779
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18760779 10 mg 10mg
18750779 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18760779 Supplier R&D Systems™ Supplier No. 4569/10
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective IKK inhibitor; orally active

PS 1145 dihydrochloride is a selective IκB kinase (IKK) inhibitor (IC50 = 100 nM). Has no effect on PKA, PKC and CKII activity. Inhibits IκB-α phosphorylation and NFκB activation in HeLa cells. Inhibits NFκB-mediated proinflammatory gene expression in human airway epithelial cells. Enhances TNF-α-induced apoptosis in a multiple myeloma cell line. Attenuates LPS-induced TNF-α production in vivo. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type PS 1145 dihydrochloride
Molecular Weight (g/mol) 395.67
Product Line Tocris
Inhibitors PS 1145 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.