missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pS2 / pNR-2 / Trefoil Factor 1 Mouse, Clone: GE2; same as R47/94, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7031-MSM1-P1
This item is not returnable.
View return policy
Specifications
| pS2 / pNR-2 / Trefoil Factor 1 | |
| Cytoplasmic | |
| GE2; same as R47/94 | |
| Unconjugated | |
| NP_003216 | |
| Mouse | |
| 6.5kDa | |
| Protein A/G Purified | |
| RUO | |
| 7031 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| TFF1 | |
| Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2. | |
| Breast or Ovarian carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction