missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAP Mouse, Clone: SPM312, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | PSAP |
|---|---|
| Clone | SPM312 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15980688
|
EnquireBio
55-MSM3X-P0 |
20 μg | |||||||
|
15990688
|
EnquireBio
55-MSM3X-P1 |
100 μg |
€ 554.40 € 478.00 / 100µg Save € 76.40 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| PSAP | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 55 | |
| Prostatic acid phosphatase purified from human seminal plasma | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 52kDa |
| SPM312 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001127666 | |
| ACPP | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic | |
| PC12 cells or normal prostate or prostate carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title