Learn More
PSAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20180-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
| PSAT1 | |
| Polyclonal | |
| Unconjugated | |
| PSAT1 | |
| EPIP, Phosphoserine aminotransferase, PSA, PSAT, PSAT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 107272, 293820, 29968 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99K85, Q9Y617 | |
| Psat1 | |
| PSAT1 Fusion Protein Ag14044 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |