Learn More
Tocris Bioscience™ PSB 069
GREENER_CHOICE
Product Code. 18085992
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18085992
Brand: Tocris Bioscience™ 2573/50
Non-selective NTPDase inhibitor
Non-selective nucleoside triphosphate diphosphohydrolase (NTPDase) inhibitor. Reported to inhibit rat NTPDases 1, 2 and 3 with similar potencies.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78510-31-3
Molecular Formula C20H12ClN2NaO5S
Specifications
Quantity 50 mg
Formula Weight 450.83
Purity >98%
Molecular Weight (g/mol) 450.825
Chemical Name or Material PSB 069