missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PSB 12379

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
PSB 12379 is a selective high affinity ecto-5'-nucleotidase/CD73 inhibitor (Ki values are 2.21 and 9.03 nM at human and rat CD73, respectively). Blocks ecto-5'-nucleotidase-mediated adenosine production by preventing the conversion of AMP to adenosine. Exhibits selectivity over other ecto-nucleotidases and ADP-activated P2Y receptors tested.
Specifications
Specifications
| Chemical Name or Material | N6-Benzyl-α,β-methyleneadenosine 5′-diphosphate disodium salt |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C18H21N5Na2O9P2 |
| Formula Weight | Observed MW: 559.31 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction