missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSCA Monoclonal Antibody (3F4E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548638
This item is not returnable.
View return policy
Description
PSCA Monoclonal Antibody for Western Blot, Flow, ELISA
PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.
Specifications
| PSCA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O43653 | |
| PSCA | |
| Purified recombinant fragment of human PSCA (AA: 1-114) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 3F4E4 | |
| Unconjugated | |
| PSCA | |
| 2210408B04Rik; caldesmon; CDM; HCAD; H-CAD; LCAD; L-CAD; NAG22; PRO232; prostate stem cell antigen; PSCA; UNQ206; UNQ206/PRO232 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8000 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction