Learn More
PSD-95 Antibody (1H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26619-50ul
 View more versions of this product
Product Code. 30043365
  • € 343.00 / 50µL
Description
PSD-95 Monoclonal antibody specifically detects PSD-95 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSD-95
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA
1H4
Western Blot 1:500-1:5000, ELISA
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15
A synthesized peptide derived from Human PSD-95 [UniProt P78352]
50 μL
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
1742
Store at -20 to -70C. Avoid freeze-thaw cycles.