missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD-95 Antibody (OTI12A5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73679PE
This item is not returnable.
View return policy
Description
PSD-95 Monoclonal antibody specifically detects PSD-95 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PSD-95 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI12A5 | |
| Western Blot | |
| discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
| Full length human recombinant protein of human DLG4 (NP_001356) produced in HEK293T cell. | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 1742 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction