missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598091
This item is not returnable.
View return policy
Description
PSD Polyclonal Antibody for Western Blot, IHC (P), ELISA
PSD, which called EFA6A, is 63% identical to EFA6B (PSD4) and contains a Sec7 domain adjacent to a PH domain, followed by a C-terminal coiled-coil motif. This gene is mapped to chromosome 10q24. And it encodes a Plekstrin homology and SEC7 domains-containing protein that functions as a guanine nucleotide exchange factor. The encoded protein regulates signal transduction by activating ADP-ribosylation factor 6. Alternative splicing results in multiple transcript variants.
Specifications
| PSD | |
| Polyclonal | |
| Unconjugated | |
| PSD | |
| 1110007H17Rik; A930015K15; EFA6; EFA6A; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ARF6; Exchange factor for ARF6 A; KIAA2011; mKIAA2011; PH and SEC7 domain-containing protein 1; pleckstrin and Sec7 domain containing; pleckstrin and Sec7 domain containing homolog; pleckstrin homology and SEC7 domain-containing protein 1; pleckstrin-Sec7 domains-containing; Psd; Psd1; Psdl; TYL | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5662 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.57 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A5PKW4 | |
| PSD | |
| Recombinant Human PH and SEC7 domain-containing protein 1 protein (247-489AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction