missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 4 Antibody (93), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08415PE
This item is not returnable.
View return policy
Description
Alpha Actinin 4 Monoclonal antibody specifically detects Alpha Actinin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Alpha Actinin 4 | |
| Monoclonal | |
| PE | |
| actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
| Recombinant full-length human Alpha Actinin 4 protein (Uniprot: O43707) | |
| 0.1 mL | |
| Phospho Specific | |
| 81 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 93 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction