missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD93 [p Tyr340] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38644-20ul
This item is not returnable.
View return policy
Description
PSD93 Polyclonal antibody specifically detects PSD93 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PSD93 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Chapsyn-110, discs, large homolog 2 (Drosophila), discs, large homolog 2, chapsyn-110, discs, large homolog 2, chapsyn-110 (Drosophila), disks large homolog 2, DKFZp781D1854, DKFZp781E0954, FLJ37266, MGC131811, Postsynaptic density protein PSD-93, PSD-93, PSD93,110kDa | |
| A synthetic phosphorylated peptide around Y340 of human PSD93 (NP_001355.2).,, Sequence:, DDYTR | |
| 20 μL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 1740 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction