missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSD93 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585780
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PSD-93, also known as chapsyn-110, is one of a family of plasma membrane-associated proteins found in synaptic junctions. PSD-93 is unique among family members in its expression in Purkinje neuron cell bodies and dendrites. PSD-93 has three ~90 amino acid repeats called PDZ domains, a single interior SH3 domain, and a carboxyl-terminal guanylate kinase homology (GuK) domain that is enzymatically inactive. It is hypothesized that PDZ-domain interactions play a role in receptor and channel clustering which contributes to neuronal plasticiyt. PSD-93 is believed to participate in the clustering of certain proteins, including NMDA receptors and shaker-type potassium channels at the synaptic membrane. There are two principal modes of interaction between PSD-93 and other proteins. NMDA receptors and shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif may interact with PSD-93 by binding to its PDZ domains. Neuronal nitric oxide synthase (nNOS), which lacks the T/SXV motif but which has its own PDZ domain, has been shown to associate with PSD-93 in vitro through a pseudo-homotypic PDZ-PDZ interaction.
Specifications
| PSD93 | |
| Polyclonal | |
| Unconjugated | |
| DLG2 | |
| A330103J02Rik; B230218P12Rik; B330007M19Rik; channel-associated protein of synapse-110; channel-associated protein of synapses, 110kDa; Chapsyn-1; chapsyn-110; discs large 2; discs large homolog 2; discs large MAGUK scaffold protein 2; discs, large homolog 2; discs, large homolog 2 (Drosophila); discs, large homolog 2, chapsyn-110; disks large homolog 2; DLG2; Dlgh2; Gm1197; Postsynaptic density protein PSD-93; PPP1R58; protein phosphatase 1, regulatory subunit 58; PSD93; PSD-93; synaptic density protein PSD-93 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23859, 64053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q63622, Q91XM9 | |
| DLG2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse PSD93. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction