missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD95 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Spezifikation
| Antigen | PSD95 |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30237126
|
Proteintech
30255-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235954
|
Proteintech
30255-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The PSD95 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PSD95. This antibody recognizes human, mouse, rat antigen. The PSD95 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Spezifikation
| PSD95 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ97752; FLJ98574; PSD95; SAP-90; SAP90 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 450 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1742 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts